Assay Details
| Gene Symbol: Qsox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042036.1 | - | 3 - 4 | 466 | 152 | |
| AK140324.1 | - | 6 - 7 | 844 | 152 | ||
| AK157992.1 | - | 6 - 7 | 837 | 152 | ||
| AK165642.1 | - | 6 - 7 | 846 | 152 | ||
| BC030934.1 | - | 6 - 7 | 878 | 4920915 | 152 | |
More Information
Updated Mapping Information
Set Membership: |
|

Qsox2
Genomic Map