Assay Details
| Gene Symbol: Qsox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042036.1 | - | 5 - 6 | 728 | 77 | |
| AK140324.1 | - | 8 - 9 | 1106 | 77 | ||
| AK157992.1 | - | 8 - 9 | 1099 | 77 | ||
| AK165642.1 | - | 8 - 9 | 1108 | 77 | ||
| BC030934.1 | - | 8 - 9 | 1140 | 4920915 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Qsox2
Genomic Map