Assay Details
| Gene Symbol: Vrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011705.3 | NP_035835.1 | 12 - 13 | 1259 | 82 | |
| GenBank mRNA | AF080253.1 | - | 12 - 13 | 1213 | 82 | |
| AK088674.1 | - | 12 - 13 | 1242 | 82 | ||
| BB749810.1 | - | 2 - 3 | 124 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vrk1
Genomic Map