Assay Details
Gene Symbol: Hspbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024172.3 | NP_077134.1 | 7 - 8 | 1161 | 102 | |
GenBank mRNA | AF338351.1 | - | 6 - 7 | 1099 | 102 | |
AK013263.1 | - | 7 - 8 | 1152 | 102 | ||
AK088469.1 | - | 7 - 8 | 1137 | 102 | ||
AK160128.1 | - | 7 - 8 | 1157 | 102 | ||
BC014758.1 | - | 7 - 8 | 1161 | 4014661 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140141 | AK088469.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140141 | AK209519.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140141 | NM_024172.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140141 | AF338351.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140141 | AK013263.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140141 | AK088469.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140141 | AK209519.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140141 | NM_024172.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140141 | AF338351.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140141 | AK013263.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627659 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717912 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186582 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557555 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 570502 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609730 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815958 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855499 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882539 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136337 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403246 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188019 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163998 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564041 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687552 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477407 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617673 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10211 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112516 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115633 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168758 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140141 | AK088469.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140141 | AK209519.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140141 | NM_024172.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140141 | AF338351.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140141 | AK013263.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140141 | AK088469.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140141 | AK209519.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140141 | NM_024172.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140141 | AF338351.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140141 | AK013263.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949974 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949969 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949986 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949970 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949971 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949967 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949882 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949891 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950002 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949977 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949992 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949884 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949892 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949988 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949973 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949978 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949985 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949898 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949982 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949881 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949993 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949888 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949990 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949890 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949894 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949998 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949897 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01475763 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168758 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115633 | - | - |
Whole Mouse Genome G4122A | A_51_P140140 | AK088469.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P140140 | AK209519.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P140140 | BC014758.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P140140 | NM_024172.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P140140 | AF338351.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138707 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103303 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114199 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168758 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115633 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05908270 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05908259 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05908272 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06195441 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06481212 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06513093 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05908271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06610560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949904 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949905 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05908260 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06513093 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05909619 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949902 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949963 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949885 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06610559 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06219703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06513094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06453067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06210420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949950 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06372895 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05992506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949881 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06610563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949898 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06524994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949897 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949891 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05908270 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949944 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949964 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949906 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949955 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05908262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06089611 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949895 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949967 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05908267 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949899 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05908271 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949882 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06638546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05908259 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06486404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949886 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949946 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06605649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949890 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949949 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10559639 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559636 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559637 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559640 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559641 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559642 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559638 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559643 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416894_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10559635 | - | - |
Mouse Genome 430 2.0 Array | 1416894_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1416894_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66245_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66245_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700019783.mm.1 | TC0700002167.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700019783.mm.1 | NM_024172.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700019783.mm.1 | NM_024172.3 | Refseq |
Mouse Expression Set 430 | 1416894_PM_at | - | - |
Mouse Expression Set 430 | 1416894_at | - | - |
Murine Genome U74v2 Set | 100987_PM_f_at | - | - |
Murine Genome U74v2 Set | 100987_f_at | - | - |
Murine Genome U74v2 Set | 167274_f_at | - | - |
Murine Genome U74v2 Set | 167255_PM_at | - | - |
Murine Genome U74v2 Set | 167274_PM_f_at | - | - |
Murine Genome U74v2 Set | 167255_at | - | - |
Mouse Genome 430A 2.0 Array | 1416894_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1416894_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416894_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416894_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 465245 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2030037 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002030037 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224137 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1224137 | - | - |
mouse-6 v1.1 (Illumina) | scl31806.5.1_30-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2030037 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl31806.5.1_30-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2030037 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2680463 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2680463 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31806.5.1_30-S | - | - |
