Assay Details
| Gene Symbol: Ppp6r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172894.2 | NP_766482.2 | 7 - 8 | 1316 | 75 | |
| GenBank mRNA | BC053076.1 | - | 7 - 8 | 1316 | 5698212 | 75 |
| BC141039.1 | - | 6 - 7 | 863 | 9055651 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp6r1
Genomic Map