Assay Details
| Gene Symbol: Acvr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111030.1 | NP_001104500.1 | 3 - 4 | 619 | 118 | |
| GenBank mRNA | AK142396.1 | - | 3 - 4 | 622 | 118 | |
| BC145224.1 | - | 2 - 3 | 415 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acvr1c
Genomic Map