Assay Details
| Gene Symbol: Acvr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033369.3 | NP_001028541.1 | 5 - 6 | 811 | 123 | |
| NM_001111030.1 | NP_001104500.1 | 6 - 7 | 1178 | 123 | ||
| GenBank mRNA | AK049089.1 | - | 5 - 6 | 812 | 123 | |
| AK142396.1 | - | 6 - 7 | 1182 | 123 | ||
| BC028780.1 | - | 2 - 3 | 196 | 123 | ||
| BC145224.1 | - | 5 - 6 | 974 | 123 | ||
More Information
Set Membership: |
|

Acvr1c
Genomic Map