Assay Details
| Gene Symbol: Olfr1112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146661.2 | NP_666872.2 | 1 | 461 | 97 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983338 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776144 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129844 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983338 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P114585 | NM_146661.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21405103 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129844 | - | - |
| Whole Mouse Genome G4122A | A_52_P114585 | NM_146661.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133994 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129844 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01215617 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02005638 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02005628 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02005636 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02005640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21405106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02005631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21405116 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10473514 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10473513 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 473493 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2900204 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002900204 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232053 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232053 | - | - |
| mouse-6 v1.1 (Illumina) | scl20673.1.139_3-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2900204 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20673.1.139_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900204 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1232053 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232053 | - | - |

Olfr1112
Genomic Map