Assay Details
| Gene Symbol: 6430503K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108091.1 | 1 - 2 | 33 | 76 | ||
| GenBank mRNA | AK020097.1 | - | 1 - 2 | 34 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
NK2 homeobox 2, opposite strand |
Nkx2-2os |
Gm14112, Nkx2-2as, Nkx2.2AS, OTTMUSG00000015691 |
Sense | Chr. 2 - 147184083 - 147331681 on NCBI Build GRCm38 | |
| Downstream | expressed sequence AI646519 | AI646519 |
Aspax1, OTTMUSG00000015701 |
173.8kb | Anti-sense | Chr. 2 - 147361541 - 147362793 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

6430503K07Rik
Genomic Map