Assay Details
Gene Symbol: Mdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012335.2 | NP_001012335.1 | 4 - 5 | 562 | 67 | |
NM_001012336.2 | NP_001012336.1 | 4 - 5 | 559 | 67 | ||
NM_001291481.1 | NP_001278410.1 | 4 - 5 | 469 | 67 | ||
NM_001291482.1 | NP_001278411.1 | 3 - 4 | 482 | 67 | ||
NM_010784.5 | NP_034914.1 | 4 - 5 | 761 | 67 | ||
GenBank mRNA | AK032742.1 | - | 4 - 5 | 465 | 67 | |
AK084457.1 | - | 4 - 5 | 482 | 67 | ||
AY864926.1 | - | 4 - 5 | 495 | 67 | ||
BC012244.1 | - | 4 - 5 | 430 | 4167496 | 67 | |
CF728526.1 | - | 4 - 5 | 455 | 67 | ||
CF738400.1 | - | 4 - 5 | 567 | 67 | ||
CF740555.1 | - | 2 - 3 | 279 | 67 | ||
CJ174578.1 | - | 3 - 4 | 357 | 67 | ||
DQ323886.1 | - | 4 - 5 | 537 | 67 | ||
M19662.1 | - | 3 - 4 | 640 | 67 | ||
M34327.1 | - | 4 - 5 | 435 | 67 | ||
M34328.1 | - | 4 - 5 | 746 | 67 | ||
M35833.1 | - | 4 - 5 | 435 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
behavioral fear response
multicellular organism development
short-term memory
response to wounding
cell migration
cerebellar granular layer development
hippocampus development
cell differentiation
adrenal gland development
defecation
negative regulation of neuron apoptotic process
positive regulation of transcription, DNA-templated
regulation of behavior
positive regulation of cell division
negative regulation of neuron death
growth factor activity
heparin binding
behavioral fear response
multicellular organism development
short-term memory
response to wounding
cell migration
cerebellar granular layer development
hippocampus development
cell differentiation
adrenal gland development
defecation
negative regulation of neuron apoptotic process
positive regulation of transcription, DNA-templated
regulation of behavior
positive regulation of cell division
negative regulation of neuron death
growth factor activity
heparin binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P107379 | M34327.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P107379 | M34328.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P107379 | M35833.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P107379 | DQ323886.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P107379 | AY864926.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971897 | M34327.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971897 | M34328.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971897 | M35833.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971897 | DQ323886.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971897 | DQ323887.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397019 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114776 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P107379 | M34327.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P107379 | M34328.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P107379 | M35833.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P107379 | DQ323886.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P107379 | AY864926.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971897 | M34327.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971897 | M34328.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971897 | M35833.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971897 | DQ323886.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971897 | DQ323887.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P453635 | - | - |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P225665 | M34327.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P225665 | M34328.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P225665 | M35833.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P225665 | DQ323886.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P225665 | DQ323887.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425467 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425474 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425458 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425441 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425430 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425445 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425453 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425439 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425469 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425456 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425428 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425448 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425433 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425449 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425465 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425460 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425476 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425463 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425470 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425435 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425429 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425434 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425462 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425426 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425447 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425437 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425450 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21425479 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114776 | - | - |
Whole Mouse Genome G4122A | A_51_P453635 | - | - |
Whole Mouse Genome G4122A | A_51_P225665 | M34327.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225665 | M34328.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225665 | M35833.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225665 | DQ323886.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P225665 | DQ323887.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112185 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114776 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01949035 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01202497 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01949042 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01949036 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01949031 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01949033 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01949041 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01202498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21425445 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21425434 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21425443 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01949041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01949031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21425441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21425433 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21425439 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21425449 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21425445 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21425435 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01949035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21425448 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21425437 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10485080 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10485078 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10485073 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10485071 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10485077 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10485079 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381602 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10485074 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10485076 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10485072 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10485075 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416006_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10381601 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10485070 | - | - |
Mouse Genome 430 2.0 Array | 1416006_at | - | - |
Mouse Genome 430 2.0 Array | 1416006_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17242_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17242_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200034581.mm.1 | NM_001012335.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034581.mm.1 | NM_001012336.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034581.mm.1 | NM_010784.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034581.mm.1 | TC0200004129.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200034582.mm.1 | NM_001012335.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034582.mm.1 | NM_001012336.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034582.mm.1 | NM_010784.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034582.mm.1 | TC0200004129.mm.1 | MTA TC |
Mouse Expression Set 430 | 1416006_at | - | - |
Mouse Expression Set 430 | 1416006_PM_at | - | - |
Murine Genome U74v2 Set | 160561_PM_at | - | - |
Murine Genome U74v2 Set | 160561_at | - | - |
Mouse Genome 430A 2.0 Array | 1416006_at | - | - |
Mouse Genome 430A 2.0 Array | 1416006_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416006_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416006_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3840020 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3840020 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003840020 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254450 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1254450 | - | - |
mouse-6 v1.1 (Illumina) | scl18991.5.1_45-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3840020 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl18991.5.1_45-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840020 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3080012 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3158499 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1254450 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3079236 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3080012 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254450 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3079236 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3158499 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl18991.5.1_45-S | - | - |
