Assay Details
| Gene Symbol: Ambra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080754.1 | NP_001074223.1 | 11 - 12 | 2511 | 62 | |
| NM_172669.3 | NP_766257.3 | 10 - 11 | 2784 | 62 | ||
| GenBank mRNA | AK084557.1 | - | 10 - 11 | 2766 | 62 | |
| AK129433.1 | - | 6 - 7 | 2048 | 62 | ||
| AK148887.1 | - | 10 - 11 | 2416 | 62 | ||
| AK155513.1 | - | 10 - 11 | 2747 | 62 | ||
| AK169280.1 | - | 11 - 12 | 2464 | 62 | ||
| BC032293.1 | - | 11 - 12 | 2434 | 62 | ||
| BC054082.1 | - | 10 - 11 | 63 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ambra1
Genomic Map