Assay Details
| Gene Symbol: Ambra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080754.1 | NP_001074223.1 | 4 - 5 | 735 | 89 | |
| NM_172669.3 | NP_766257.3 | 4 - 5 | 735 | 89 | ||
| GenBank mRNA | AK037516.1 | - | 4 - 5 | 692 | 89 | |
| AK083596.1 | - | 4 - 5 | 692 | 89 | ||
| AK084557.1 | - | 4 - 5 | 716 | 89 | ||
| AK148887.1 | - | 4 - 5 | 727 | 89 | ||
| AK169280.1 | - | 4 - 5 | 688 | 89 | ||
| BC032293.1 | - | 4 - 5 | 658 | 89 | ||
| BC054082.1 | - | 4 - 5 | 667 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ambra1
Genomic Map