Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200009066.mm.1 |
NM_007421.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009066.mm.1 |
NM_007421.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009066.mm.1 |
TC1200001262.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009068.mm.1 |
NM_007421.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009068.mm.1 |
NM_007421.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009068.mm.1 |
TC1200001262.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009062.mm.1 |
NM_007421.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009062.mm.1 |
NM_007421.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009062.mm.1 |
TC1200001262.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009065.mm.1 |
NM_007421.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009065.mm.1 |
NM_007421.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009065.mm.1 |
TC1200001262.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009063.mm.1 |
NM_007421.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009063.mm.1 |
NM_007421.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009063.mm.1 |
TC1200001262.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009064.mm.1 |
NM_007421.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009064.mm.1 |
NM_007421.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009064.mm.1 |
TC1200001262.mm.1 |
MTA TC |