Assay Details
Gene Symbol: Lrrc75aos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019771.1 | - | 2 - 3 | 286 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | leucine rich repeat containing 75A | Lrrc75a | Fam211a | Anti-sense | Chr. 11 - 62604884 - 62648583 on NCBI Build GRCm38 | |
Downstream | membrane magnesium transporter 2 | Mmgt2 | AI852046 | 11.0kb | Sense | Chr. 11 - 62648632 - 62666359 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
