Assay Details
Gene Symbol: 8030411F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030135.2 | NP_084411.1 | 2 - 3 | 414 | 71 | |
GenBank mRNA | AF440735.1 | - | 2 - 3 | 414 | 71 | |
AK020193.1 | - | 2 - 3 | 330 | 71 | ||
BC132218.1 | - | 2 - 3 | 359 | 40130495 | 71 | |
BC132222.1 | - | 2 - 3 | 359 | 40130499 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000598 | AF440735.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000598 | AK020193.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2000598 | NM_030135.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808273 | AF102524.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808273 | BC127978.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808273 | NM_146946.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180274 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106907 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000598 | AF440735.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000598 | AK020193.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2000598 | NM_030135.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732163 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732159 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732151 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732157 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732154 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732166 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732155 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732165 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732160 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732152 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732150 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732164 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106907 | - | - |
Whole Mouse Genome G4122A | A_51_P426837 | AF440735.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426837 | AK020193.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426837 | BC132218.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426837 | NM_030135.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P426837 | BC132222.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106907 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01972466 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01972438 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01972465 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01972437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732213 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732122 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01972437 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01450396 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732165 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732133 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01766800 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732176 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732173 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732123 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01972436 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732141 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732155 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732129 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732137 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732189 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732131 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732175 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01268819 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732182 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732159 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732168 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732143 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732125 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01972464 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01972438 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01164250 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732185 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02126907 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01766801 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732174 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02126909 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01972441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01408251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01996934 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732179 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732186 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01972466 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732157 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732144 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732147 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732163 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732134 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732160 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732124 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21732126 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10476908 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10476907 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10476909 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451712_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10476906 | - | - |
Mouse Genome 430 2.0 Array | 1451712_at | - | - |
Mouse Genome 430 2.0 Array | 1451712_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78609_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78609_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200017402.mm.1 | NM_030135.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017402.mm.1 | NM_030135.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017402.mm.1 | TC0200002199.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200017404.mm.1 | NM_030135.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017404.mm.1 | NM_030135.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017404.mm.1 | TC0200002199.mm.1 | MTA TC |
Mouse Expression Set 430 | 1451712_at | - | - |
Mouse Expression Set 430 | 1451712_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451712_at | - | - |
Mouse Genome 430A 2.0 Array | 1451712_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451712_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451712_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 692595 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2636508 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2636508 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2636508 | - | - |
