Assay Details
| Gene Symbol: Syt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030725.4 | NP_109650.1 | 3 - 4 | 635 | 76 | |
| GenBank mRNA | AB048947.1 | - | 3 - 4 | 561 | 76 | |
| AK044038.1 | - | 3 - 4 | 639 | 76 | ||
| AK049840.1 | - | 3 - 4 | 622 | 76 | ||
| AK082425.1 | - | 3 - 4 | 636 | 76 | ||
| AK083803.1 | - | 3 - 4 | 636 | 76 | ||
| BC030907.1 | - | 3 - 4 | 625 | 4503633 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syt13
Genomic Map