Assay Details
| Gene Symbol: Lgals8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199043.1 | NP_001185972.1 | 4 - 5 | 938 | 72 | |
| NM_001291055.1 | NP_001277984.1 | 3 - 4 | 440 | 72 | ||
| NM_001291057.1 | NP_001277986.1 | 3 - 4 | 663 | 72 | ||
| NM_001291060.1 | NP_001277989.1 | 3 - 4 | 609 | 72 | ||
| NM_018886.5 | NP_061374.1 | 4 - 5 | 752 | 72 | ||
| GenBank mRNA | AF218069.1 | - | 3 - 4 | 454 | 72 | |
| AK004782.2 | - | 4 - 5 | 620 | 72 | ||
| AK029526.1 | - | 5 - 6 | 800 | 72 | ||
| AK029685.1 | - | 4 - 5 | 647 | 72 | ||
| AK075736.1 | - | 3 - 4 | 419 | 72 | ||
| AK083350.1 | - | 4 - 5 | 505 | 72 | ||
| AK145881.1 | - | 4 - 5 | 472 | 72 | ||
| AK146746.1 | - | 4 - 5 | 481 | 72 | ||
| AK155066.1 | - | 3 - 4 | 378 | 72 | ||
| BC040243.1 | - | 4 - 5 | 586 | 2582159 | 72 | |
| CT010251.1 | - | 3 - 4 | 339 | 72 | ||
| EF524570.1 | - | 3 - 4 | 339 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lgals8
Genomic Map