Assay Details
| Gene Symbol: Lgals8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199043.1 | NP_001185972.1 | 5 - 6 | 1063 | 63 | |
| NM_001291057.1 | NP_001277986.1 | 4 - 5 | 788 | 63 | ||
| NM_001291060.1 | NP_001277989.1 | 4 - 5 | 734 | 63 | ||
| NM_018886.5 | NP_061374.1 | 5 - 6 | 877 | 63 | ||
| GenBank mRNA | AF218069.1 | - | 4 - 5 | 579 | 63 | |
| AK004782.2 | - | 5 - 6 | 745 | 63 | ||
| AK029526.1 | - | 6 - 7 | 925 | 63 | ||
| AK029685.1 | - | 5 - 6 | 772 | 63 | ||
| AK075736.1 | - | 4 - 5 | 544 | 63 | ||
| AK083350.1 | - | 5 - 6 | 630 | 63 | ||
| AK145881.1 | - | 5 - 6 | 597 | 63 | ||
| AK146746.1 | - | 5 - 6 | 606 | 63 | ||
| AK148939.1 | - | 5 - 6 | 643 | 63 | ||
| AK155066.1 | - | 4 - 5 | 503 | 63 | ||
| AK199736.1 | - | 1 - 2 | 73 | 63 | ||
| BC040243.1 | - | 5 - 6 | 711 | 2582159 | 63 | |
| CT010251.1 | - | 4 - 5 | 464 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lgals8
Genomic Map