Assay Details
| Gene Symbol: Pak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195046.1 | NP_001181975.1 | 13 - 14 | 1345 | 64 | |
| NM_001195047.1 | NP_001181976.1 | 12 - 13 | 1300 | 64 | ||
| NM_001195048.1 | NP_001181977.1 | 13 - 14 | 1455 | 64 | ||
| NM_001195049.1 | NP_001181978.1 | 15 - 16 | 1719 | 64 | ||
| NM_008778.3 | NP_032804.2 | 12 - 13 | 1328 | 64 | ||
| GenBank mRNA | AF082297.1 | - | 11 - 12 | 1242 | 64 | |
| AJ496262.1 | - | 11 - 12 | 1210 | 64 | ||
| AJ496263.1 | - | 12 - 13 | 1255 | 64 | ||
| AK031853.1 | - | 13 - 14 | 1336 | 64 | ||
| AK158893.1 | - | 13 - 14 | 1455 | 64 | ||
| AM943853.1 | - | 12 - 13 | 1273 | 64 | ||
| AM943854.1 | - | 13 - 14 | 1318 | 64 | ||
| BC053403.1 | - | 12 - 13 | 1335 | 30060082 | 64 | |
| U39738.1 | - | 16 - 17 | 1726 | 64 | ||
More Information
Set Membership: |
|

Pak3
Genomic Map