| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00019554.mm.1 |
NM_001114385.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019554.mm.1 |
NM_001114385.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019554.mm.1 |
NM_031258.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019554.mm.1 |
NM_031258.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019554.mm.1 |
TC0X00003113.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00019553.mm.1 |
NM_001114385.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019553.mm.1 |
NM_001114385.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019553.mm.1 |
NM_031258.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019553.mm.1 |
NM_031258.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019553.mm.1 |
TC0X00003113.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00019551.mm.1 |
NM_001114385.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019551.mm.1 |
NM_001114385.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019551.mm.1 |
NM_031258.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019551.mm.1 |
NM_031258.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019551.mm.1 |
TC0X00003113.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00019552.mm.1 |
NM_001114385.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019552.mm.1 |
NM_001114385.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019552.mm.1 |
NM_031258.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019552.mm.1 |
NM_031258.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019552.mm.1 |
TC0X00003113.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00019555.mm.1 |
NM_001114385.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019555.mm.1 |
NM_001114385.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019555.mm.1 |
NM_031258.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019555.mm.1 |
NM_031258.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019555.mm.1 |
TC0X00003113.mm.1 |
MTA TC |