Assay Details
| Gene Symbol: Chrdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114385.1 | NP_001107857.1 | 2 - 3 | 606 | 169 | |
| NM_031258.3 | NP_112548.2 | 2 - 3 | 606 | 169 | ||
| GenBank mRNA | AF296451.1 | - | 2 - 3 | 528 | 169 | |
| AF305714.1 | - | 1 - 2 | 145 | 169 | ||
| AK142805.1 | - | 2 - 3 | 230 | 169 | ||
| AK164661.1 | - | 2 - 3 | 201 | 169 | ||
| BC066832.1 | - | 2 - 3 | 240 | 30102312 | 169 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF296451.1, AF305714.1, AK142805.1, AK164661.1, BC066832.1, NM_001114385.1, NM_031258.3 transcripts.
Set Membership: |
|

Chrdl1
Genomic Map