Assay Details
| Gene Symbol: Chrdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114385.1 | NP_001107857.1 | 3 - 4 | 722 | 79 | |
| NM_031258.3 | NP_112548.2 | 3 - 4 | 722 | 79 | ||
| GenBank mRNA | AF296451.1 | - | 3 - 4 | 644 | 79 | |
| AF305714.1 | - | 2 - 3 | 261 | 79 | ||
| AF321853.1 | - | 2 - 3 | 198 | 79 | ||
| AK142805.1 | - | 3 - 4 | 346 | 79 | ||
| AK164661.1 | - | 3 - 4 | 317 | 79 | ||
| BC050818.1 | - | 2 - 3 | 212 | 79 | ||
| BC066832.1 | - | 3 - 4 | 356 | 30102312 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chrdl1
Genomic Map