Assay Details
| Gene Symbol: Lair1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113474.1 | NP_001106945.1 | 4 - 5 | 619 | 57 | |
| NM_178611.6 | NP_848726.1 | 3 - 4 | 289 | 57 | ||
| GenBank mRNA | AF479685.1 | - | 4 - 5 | 554 | 57 | |
| AF491829.1 | - | 4 - 5 | 652 | 57 | ||
| AF491830.1 | - | 3 - 4 | 201 | 57 | ||
| AF491831.1 | - | 4 - 5 | 531 | 57 | ||
| AK033472.1 | - | 3 - 4 | 266 | 57 | ||
| AK038574.1 | - | 3 - 4 | 252 | 57 | ||
| AK043664.1 | - | 3 - 4 | 346 | 57 | ||
| AK044465.1 | - | 4 - 5 | 590 | 57 | ||
| AK050281.1 | - | 4 - 5 | 575 | 57 | ||
| AK054158.1 | - | 4 - 5 | 698 | 57 | ||
| AY392763.1 | - | 4 - 5 | 556 | 57 | ||
| AY392764.1 | - | 3 - 4 | 226 | 57 | ||
| BC025901.1 | - | 4 - 5 | 583 | 5253846 | 57 | |
| BY204263.1 | - | 2 - 3 | 233 | 57 | ||
More Information
Set Membership: |
|

Lair1
Genomic Map