Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200027800.mm.1 |
NM_022415.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027800.mm.1 |
NM_022415.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027800.mm.1 |
TC0200003278.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200027801.mm.1 |
NM_022415.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027801.mm.1 |
NM_022415.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027801.mm.1 |
TC0200003278.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200027798.mm.1 |
NM_022415.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027798.mm.1 |
NM_022415.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027798.mm.1 |
TC0200003278.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200027802.mm.1 |
NM_022415.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027802.mm.1 |
NM_022415.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027802.mm.1 |
TC0200003278.mm.1 |
MTA TC |