Assay Details
| Gene Symbol: Fcnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010190.1 | NP_034320.1 | 6 - 7 | 447 | 78 | |
| GenBank mRNA | AF063217.1 | - | 6 - 7 | 420 | 78 | |
| AK010913.1 | - | 5 - 6 | 445 | 78 | ||
| AK151676.1 | - | 6 - 7 | 455 | 78 | ||
| BC107220.1 | - | 6 - 7 | 447 | 40051808 | 78 | |
| BC107221.1 | - | 6 - 7 | 447 | 40051810 | 78 | |
More Information
Set Membership: |
|

Fcnb
Genomic Map