Assay Details
| Gene Symbol: Wdr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080848.2 | NP_543124.1 | 10 - 11 | 921 | 78 | |
| GenBank mRNA | AF416510.1 | - | 10 - 11 | 926 | 78 | |
| AK075937.1 | - | 10 - 11 | 864 | 78 | ||
| AK144091.1 | - | 10 - 11 | 865 | 78 | ||
| BC008547.1 | - | 5 - 6 | 301 | 78 | ||
| BC016103.1 | - | 10 - 11 | 856 | 4919781 | 78 | |
| BC025801.1 | - | 10 - 11 | 898 | 5132525 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr5
Genomic Map