Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600004292.mm.1 |
TC0600000560.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004292.mm.1 |
NM_001079901.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004292.mm.1 |
NM_001079901.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004292.mm.1 |
NM_001079902.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004292.mm.1 |
NM_001079902.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004292.mm.1 |
NM_001079903.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004292.mm.1 |
NM_001079903.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004292.mm.1 |
NM_001079904.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004292.mm.1 |
NM_001079904.1 |
Refseq |