Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600004594.mm.1 |
TC0600000598.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004594.mm.1 |
NR_015581.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004594.mm.1 |
NR_015581.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004590.mm.1 |
TC0600000598.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004590.mm.1 |
NR_015581.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004590.mm.1 |
NR_015581.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004590.mm.1 |
NR_027892.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004590.mm.1 |
NR_027892.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004592.mm.1 |
TC0600000598.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004592.mm.1 |
NR_015581.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004592.mm.1 |
NR_015581.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004592.mm.1 |
NR_027892.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004592.mm.1 |
NR_027892.1 |
Refseq |