Assay Details
| Gene Symbol: Syk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198977.1 | NP_001185906.1 | 2 - 3 | 625 | 78 | |
| NM_011518.2 | NP_035648.2 | 2 - 3 | 629 | 78 | ||
| GenBank mRNA | AK036736.1 | - | 2 - 3 | 553 | 78 | |
| AK049207.1 | - | 3 - 4 | 723 | 78 | ||
| AK136363.1 | - | 2 - 3 | 628 | 78 | ||
| AK143564.1 | - | 2 - 3 | 658 | 78 | ||
| BB629720.1 | - | 2 - 3 | 551 | 78 | ||
| BC065121.1 | - | 2 - 3 | 561 | 30533300 | 78 | |
| U25685.1 | - | 1 - 2 | 413 | 78 | ||
| U36776.1 | - | 2 - 3 | 632 | 78 | ||
| Z49877.1 | - | 2 - 3 | 890 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syk
Genomic Map