Assay Details
| Gene Symbol: Cst7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009977.3 | NP_034107.2 | 2 - 3 | 345 | 72 | |
| GenBank mRNA | AB015224.1 | - | 2 - 3 | 326 | 72 | |
| AF031825.1 | - | 2 - 3 | 274 | 72 | ||
| AK004420.1 | - | 2 - 3 | 346 | 72 | ||
| AK170583.1 | - | 2 - 3 | 292 | 72 | ||
| BC117047.1 | - | 2 - 3 | 330 | 40126366 | 72 | |
| BC117049.1 | - | 2 - 3 | 330 | 40126368 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cst7
Genomic Map