Assay Details
| Gene Symbol: Lce1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025420.2 | NP_079696.1 | 1 - 2 | 44 | 69 | |
| GenBank mRNA | AK019517.1 | - | 1 - 2 | 44 | 69 | |
| BC109182.1 | - | 1 - 2 | 46 | 40049867 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK019517.1, BC109182.1, NM_025420.2 transcripts.
Set Membership: |
|

Lce1m
Genomic Map