Assay Details
| Gene Symbol: Hspa14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015765.2 | NP_056580.2 | 4 - 5 | 390 | 129 | |
| GenBank mRNA | AK031489.1 | - | 329 | 129 | ||
| AK145937.1 | - | 4 - 5 | 344 | 129 | ||
| AK153397.1 | - | 4 - 5 | 352 | 129 | ||
| BC002056.1 | - | 4 - 5 | 318 | 3493119 | 129 | |
| BU505607.1 | - | 1 - 2 | 41 | 129 | ||
| U08215.1 | - | 4 - 5 | 350 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hspa14
Genomic Map