Assay Details
Gene Symbol: Olah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145921.1 | NP_666033.1 | 1 - 2 | 127 | 65 | |
GenBank mRNA | AK136031.1 | - | 2 - 3 | 383 | 65 | |
BC025001.1 | - | 1 - 2 | 127 | 5252879 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK136031.1, BC025001.1, NM_145921.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
