Assay Details
Gene Symbol: Olah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145921.1 | NP_666033.1 | 6 - 7 | 865 | 71 | |
GenBank mRNA | AK087571.1 | - | 6 - 7 | 885 | 71 | |
AK136031.1 | - | 7 - 8 | 1121 | 71 | ||
BC025001.1 | - | 6 - 7 | 865 | 5252879 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
