Assay Details
| Gene Symbol: Ucma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113558.2 | NP_001107030.1 | 2 - 3 | 251 | 95 | |
| NM_001165932.1 | NP_001159404.1 | 2 - 3 | 251 | 95 | ||
| GenBank mRNA | BC115608.2 | - | 2 - 3 | 154 | 40102702 | 95 |
| BC115609.2 | - | 2 - 3 | 154 | 40102703 | 95 | |
| BE648935.1 | - | 2 - 3 | 214 | 95 | ||
| EF529510.1 | - | 2 - 3 | 222 | 95 | ||
| EU368184.1 | - | 2 - 3 | 251 | 95 | ||
| FJ217397.1 | - | 2 - 3 | 189 | 95 | ||
| FJ217399.1 | - | 2 - 3 | 189 | 95 | ||
More Information
Set Membership: |
|

Ucma
Genomic Map