Assay Details
| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009261.3 | NP_033287.2 | 16 - 17 | 2105 | 120 | |
| GenBank mRNA | AK006314.1 | - | 3 - 4 | 288 | 120 | |
| AK078956.1 | - | 7 - 8 | 896 | 120 | ||
| BC014710.1 | - | 17 - 18 | 2189 | 4502840 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Strbp
Genomic Map