Assay Details
| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009261.3 | NP_033287.2 | 8 - 9 | 1104 | 123 | |
| GenBank mRNA | BC014710.1 | - | 9 - 10 | 1188 | 4502840 | 123 |
| X84692.1 | - | 1041 | 123 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Strbp
Genomic Map