Assay Details
| Gene Symbol: Ggta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF297615.1 | - | 4 - 5 | 93 | ||
| AK004753.1 | - | 4 - 5 | 641 | 93 | ||
| M85153.1 | - | 4 - 5 | 93 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ggta1| Gene Symbol: Ggta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF297615.1 | - | 4 - 5 | 93 | ||
| AK004753.1 | - | 4 - 5 | 641 | 93 | ||
| M85153.1 | - | 4 - 5 | 93 | |||
Set Membership: |
|