Assay Details
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163434.1 | NP_001156906.1 | 1 - 2 | 52 | 65 | |
GenBank mRNA | AK076079.1 | - | 1 - 2 | 48 | 65 | |
AK077463.1 | - | 1 - 2 | 52 | 65 | ||
AK144650.1 | - | 1 - 2 | 48 | 65 | ||
AK144742.1 | - | 1 - 2 | 48 | 65 | ||
AK146647.1 | - | 1 - 2 | 51 | 65 | ||
AK148539.1 | - | 1 - 2 | 51 | 65 | ||
AK149447.1 | - | 1 - 2 | 51 | 65 | ||
AK151647.1 | - | 1 - 2 | 50 | 65 | ||
AK151806.1 | - | 1 - 2 | 51 | 65 | ||
AK152020.1 | - | 1 - 2 | 48 | 65 | ||
AK152958.1 | - | 1 - 2 | 51 | 65 | ||
AK159719.1 | - | 1 - 2 | 48 | 65 | ||
AK160821.1 | - | 52 | 65 | |||
AK166739.1 | - | 1 - 2 | 48 | 65 | ||
AK167011.1 | - | 1 - 2 | 48 | 65 | ||
AK168012.1 | - | 1 - 2 | 52 | 65 | ||
AK168230.1 | - | 1 - 2 | 47 | 65 | ||
AK169034.1 | - | 1 - 2 | 52 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK076079.1, AK146647.1, AK148539.1, AK149447.1, AK151647.1, AK151806.1, AK152020.1, AK152958.1, AK159719.1, AK166739.1, AK167011.1, AK168012.1, AK169034.1, NM_001163434.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
