Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200005885.mm.1 |
NM_001001602.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005885.mm.1 |
NM_001001602.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005885.mm.1 |
NM_001114124.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005885.mm.1 |
NM_001114125.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005885.mm.1 |
NM_001114125.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005885.mm.1 |
TC0200000641.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200005886.mm.1 |
NM_001001602.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005886.mm.1 |
NM_001001602.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005886.mm.1 |
NM_001114124.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005886.mm.1 |
NM_001114125.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005886.mm.1 |
NM_001114125.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005886.mm.1 |
TC0200000641.mm.1 |
MTA TC |