Assay Details
Gene Symbol: Pter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK216138.1 | - | 1 - 2 | 39 | 180 | |
BB854955.1 | - | 1 - 2 | 127 | 180 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
