Assay Details
| Gene Symbol: Cacnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252533.1 | NP_001239462.1 | 4 - 5 | 596 | 75 | |
| NM_023116.4 | NP_075605.1 | 5 - 6 | 727 | 75 | ||
| NR_045533.1 | 4 - 5 | 671 | 75 | |||
| GenBank mRNA | AK020806.1 | - | 4 - 5 | 672 | 75 | |
| AK034054.1 | - | 5 - 6 | 728 | 75 | ||
| AK078220.1 | - | 4 - 5 | 452 | 75 | ||
| AM259383.2 | - | 4 - 5 | 596 | 75 | ||
| BC109156.1 | - | 5 - 6 | 659 | 40047524 | 75 | |
| BC109157.1 | - | 5 - 6 | 659 | 40047531 | 75 | |
| FM872406.1 | - | 4 - 5 | 552 | 75 | ||
| FM872407.1 | - | 4 - 5 | 658 | 75 | ||
| FM872408.1 | - | 5 - 6 | 726 | 75 | ||
| KJ789960.1 | - | 4 - 5 | 429 | 75 | ||
| L20343.1 | - | 4 - 5 | 506 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cacnb2
Genomic Map