Assay Details
| Gene Symbol: Ntng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB052337.1 | - | 3 - 4 | 1030 | 96 | |
| AF475080.1 | - | 3 - 4 | 1030 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ntng2| Gene Symbol: Ntng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB052337.1 | - | 3 - 4 | 1030 | 96 | |
| AF475080.1 | - | 3 - 4 | 1030 | 96 | ||
Set Membership: |
|