Assay Details
| Gene Symbol: 9230102O04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040511.1 | 1 - 2 | 330 | 114 | ||
| GenBank mRNA | AK020309.1 | - | 1 - 2 | 331 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GATA binding protein 3 | Gata3 | Gata-3, jal | Sense | Chr. 2 - 9857078 - 9887950 on NCBI Build GRCm38 | |
| Downstream | predicted gene 13262 | Gm13262 | OTTMUSG00000011195 | 16.6kb | Anti-sense | Chr. 2 - 9906165 - 9918096 on NCBI Build GRCm38 | 
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay targets the same location as MTA transcript cluster(s) TC0200002935.mm.1
					            	This assay targets the same location as MTA transcript cluster(s) TC0200002935.mm.1| Set Membership: |  Amplicon length greater than or equal to 101  Mus musculus  Probe spans exons | 
Gene Ontology Categories:
MicroArray Probes:
 Back To Top
 Back To Top
 9230102O04Rik
9230102O04Rik Genomic Map
Genomic Map Back To Top
 Back To Top