Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200002786.mm.1 |
NM_001199146.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002786.mm.1 |
NM_001199146.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002786.mm.1 |
NM_153410.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002786.mm.1 |
NM_153410.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002786.mm.1 |
TC0200000445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002785.mm.1 |
NM_001199146.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002785.mm.1 |
NM_001199146.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002785.mm.1 |
NM_153410.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002785.mm.1 |
NM_153410.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002785.mm.1 |
TC0200000445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002789.mm.1 |
NM_001199146.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002789.mm.1 |
NM_001199146.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002789.mm.1 |
NM_001199147.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002789.mm.1 |
NM_001199147.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002789.mm.1 |
NM_153410.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002789.mm.1 |
NM_153410.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002789.mm.1 |
TC0200000445.mm.1 |
MTA TC |