Assay Details
| Gene Symbol: Noxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163626.1 | NP_001157098.1 | 8 - 9 | 1077 | 91 | |
| NM_172204.4 | NP_757341.3 | 8 - 9 | 1101 | 91 | ||
| GenBank mRNA | AF539798.1 | - | 8 - 9 | 820 | 91 | |
| AK087263.1 | - | 8 - 9 | 1003 | 91 | ||
| AY255770.1 | - | 8 - 9 | 820 | 91 | ||
| BC047532.1 | - | 8 - 9 | 1092 | 4988389 | 91 | |
More Information
Set Membership: |
|

Noxa1
Genomic Map