Assay Details
| Gene Symbol: Gabrb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038701.2 | NP_001033790.1 | 1 - 2 | 132 | 105 | |
| GenBank mRNA | AK083107.1 | - | 1 - 2 | 133 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gabrb3
Genomic Map