Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700019817.mm.1 |
TC0700002169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019817.mm.1 |
NM_001289848.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019817.mm.1 |
NM_183406.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019813.mm.1 |
TC0700002169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019813.mm.1 |
NM_001289848.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019813.mm.1 |
NM_001289849.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019813.mm.1 |
NM_001289850.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019813.mm.1 |
NM_183405.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019813.mm.1 |
NM_183406.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019813.mm.1 |
NR_110411.1 |
Refseq |