Assay Details
| Gene Symbol: Chn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113246.2 | NP_001106717.2 | 2 - 3 | 369 | 85 | |
| NM_001166603.1 | NP_001160075.1 | 2 - 3 | 369 | 85 | ||
| GenBank mRNA | AB264772.1 | - | 2 - 3 | 66 | 85 | |
| BC054770.1 | - | 2 - 3 | 121 | 6415204 | 85 | |
| CA749754.1 | - | 2 - 3 | 369 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chn1
Genomic Map