Assay Details
| Gene Symbol: Chn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166604.1 | NP_001160076.1 | 1 - 2 | 742 | 105 | |
| NM_175752.3 | NP_786928.2 | 1 - 2 | 742 | 105 | ||
| GenBank mRNA | AB264771.1 | - | 1 - 2 | 566 | 105 | |
| AF332069.1 | - | 1 - 2 | 250 | 105 | ||
| AF332070.1 | - | 1 - 2 | 250 | 105 | ||
| AK032630.1 | - | 1 - 2 | 700 | 105 | ||
| AK049943.1 | - | 1 - 2 | 686 | 105 | ||
| AK075606.1 | - | 1 - 2 | 704 | 105 | ||
| AK147686.1 | - | 1 - 2 | 686 | 105 | ||
| BC010825.1 | - | 1 - 2 | 717 | 4219273 | 105 | |
| BC024796.1 | - | 1 - 2 | 693 | 5360438 | 105 | |
| BC025023.1 | - | 1 - 2 | 673 | 5364554 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chn1
Genomic Map